Assay Details
Target Gene Details
Entrez Gene ID: | 14756 |
Gene Name: | glycosylphosphatidylinositol specific phospholipase D1 |
Gene Aliases: |
6330541J12Rik, AW546131 |
Location: |
Chr.13:24943152-24991936 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gpld1 | NM_008156.2 | NP_032182.2 | ||
AF050666.1 | AAC77799.1 | |||
AK004957.2 | BAB23698.1 | |||
AY081194.1 | AAL87452.1 | |||
AY333960.1 | AAP93132.1 | |||
BC019146.1 | AAH19146.1 |
More Information
Set Membership: |
Intragenic Non-exonic |