Assay Details
Target Gene Details
Entrez Gene ID: | 211446 |
Gene Name: | exocyst complex component 3 |
Gene Aliases: |
2810050O03Rik, E430013E20Rik, Sec6, Sec6l1 |
Location: |
Chr.13:74169796-74208729 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Exoc3 | NM_177333.3 | NP_796307.2 | ||
AK088333.1 | BAC40288.1 | |||
AK089979.1 | ||||
AK131141.1 | BAD21391.1 | |||
AK141537.1 | BAE24723.1 | |||
BC031743.1 | AAH31743.1 |
More Information
Set Membership: |
Intragenic Non-exonic |