Assay Details
Target Gene Details
Entrez Gene ID: | 12919 |
Gene Name: | corticotropin releasing hormone binding protein |
Gene Aliases: |
CRH-BP |
Location: |
Chr.13:95431376-95444831 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crhbp | NM_198408.3 | 3 | 411 | NP_940800.1 |
AK135067.1 | 3 | 412 | BAE22406.1 | |
AK139404.1 | 3 | 257 | ||
AK157912.1 | 1 | 961 | ||
BC061247.1 | 3 | 352 | AAH61247.1 |