Assay Details
Target Gene Details
Entrez Gene ID: | 16398 |
Gene Name: | integrin alpha 2 |
Gene Aliases: |
CD49B, DX5 |
Location: |
Chr.13:114835912-114932041 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga2 | NM_008396.2 | NP_032422.2 | ||
AK036572.1 | BAC29485.1 | |||
AK051050.1 | ||||
AK156604.1 | ||||
BC065139.1 | AAH65139.1 | |||
Z29987.1 | CAA82877.1 |
More Information
Set Membership: |
Intragenic Non-exonic |