Assay Details
Target Gene Details
Entrez Gene ID: | 16398 |
Gene Name: | integrin alpha 2 |
Gene Aliases: |
CD49B, DX5 |
Location: |
Chr.13:114835912-114932041 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga2 | NM_008396.2 | 12 | 1530 | NP_032422.2 |
AK051050.1 | 12 | 1541 | ||
AK156604.1 | 12 | 1572 | ||
BC065139.1 | 12 | 1530 | AAH65139.1 | |
X75427.1 | 1 | 75 | CAA53178.1 | |
Z29987.1 | 12 | 1534 | CAA82877.1 |
More Information
Set Membership: |
Intragenic Exonic |