Assay Details
Target Gene Details
Entrez Gene ID: | 26385 |
Gene Name: | G protein-coupled receptor kinase 6 |
Gene Aliases: |
Gprk6 |
Location: |
Chr.13:55445072-55460927 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grk6 | NM_001038018.4 | NP_001033107.1 | ||
NM_001112711.2 | NP_001106182.1 | |||
NM_001286063.1 | NP_001272992.1 | |||
NM_001286064.1 | NP_001272993.1 | |||
NM_001286065.1 | NP_001272994.1 | |||
NM_001286066.1 | NP_001272995.1 | |||
NM_011938.4 | NP_036068.2 | |||
AF040747.1 | AAC09268.1 | |||
AF040748.1 | AAC09269.1 | |||
AF040749.1 | AAC09270.1 | |||
AK051405.1 | ||||
AK146119.1 | BAE26914.1 | |||
AK161142.1 | BAE36212.1 | |||
AK179644.1 | ||||
AK187852.1 | ||||
BC075719.1 | AAH75719.1 | |||
Y15797.1 | CAA75788.1 | |||
Y15798.1 | CAA75789.1 | |||
Y15799.1 | CAA75790.1 | |||
Y15800.1 | CAA75791.1 |
More Information
Set Membership: |
Intragenic Non-exonic |