Assay Details
Target Gene Details
Entrez Gene ID: | 26385 |
Gene Name: | G protein-coupled receptor kinase 6 |
Gene Aliases: |
Gprk6 |
Location: |
Chr.13:55445072-55460927 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grk6 | NM_001038018.4 | 14 | 1655 | NP_001033107.1 |
NM_001112711.2 | 14 | 1655 | NP_001106182.1 | |
NM_001286063.1 | 14 | 1635 | NP_001272992.1 | |
NM_001286064.1 | 14 | 1640 | NP_001272993.1 | |
NM_001286065.1 | 14 | 1553 | NP_001272994.1 | |
NM_001286066.1 | 14 | 1648 | NP_001272995.1 | |
NM_011938.4 | 14 | 1655 | NP_036068.2 | |
AF040747.1 | 14 | 1525 | AAC09268.1 | |
AF040748.1 | 14 | 1525 | AAC09269.1 | |
AF040749.1 | 14 | 1525 | AAC09270.1 | |
AK051405.1 | 14 | 1637 | ||
AK146119.1 | 14 | 1428 | BAE26914.1 | |
AK161142.1 | 12 | 1287 | BAE36212.1 | |
AK180303.1 | 2 | 115 | ||
BC075719.1 | 14 | 1536 | AAH75719.1 | |
CA528447.1 | 3 | 254 | ||
CD354984.1 | ||||
Y15797.1 | 14 | 1477 | CAA75788.1 | |
Y15798.1 | 14 | 1499 | CAA75789.1 | |
Y15799.1 | 14 | 1482 | CAA75790.1 | |
Y15800.1 | 14 | 1470 | CAA75791.1 |
More Information
Set Membership: |
Intragenic Exonic |