Assay Details
Target Gene Details
Entrez Gene ID: | 218138 |
Gene Name: | GDP-mannose 4, 6-dehydratase |
Gene Aliases: |
BC031788, C87208 |
Location: |
Chr.13:31819583-32338725 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gmds | NM_146041.2 | NP_666153.1 | ||
AK086076.1 | 7 | 848 | ||
BC031788.1 | AAH31788.1 | |||
BC093502.1 | AAH93502.1 |
More Information
Set Membership: |
Intragenic Non-exonic |