Assay Details
Target Gene Details
Entrez Gene ID: | 218461 |
Gene Name: | phosphodiesterase 8B |
Gene Aliases: |
B230331L10Rik, C030047E14Rik |
Location: |
Chr.13:95024105-95250397 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pde8b | NM_172263.2 | 1 | 50 | NP_758467.2 |
BC032286.1 | 1 | 62 | AAH32286.1 | |
BY244338.1 | 1 | 83 | ||
CD806392.1 |
More Information
Set Membership: |
Intragenic Exonic |