Assay Details
Target Gene Details
Entrez Gene ID: | 109700 |
Gene Name: | integrin alpha 1 |
Gene Aliases: |
CD49A, E130012M19Rik, Vla1 |
Location: |
Chr.13:114958079-115101964 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga1 | NM_001033228.3 | 26 | 3668 | NP_001028400.2 |
AK035870.1 | 26 | 3669 | BAE20498.1 | |
AK040830.1 | 17 | 2092 | ||
AK053377.1 | 1 | 67 | BAC35370.1 |
More Information
Set Membership: |
Intragenic Exonic |