Assay Details
Target Gene Details
Entrez Gene ID: | 18105 |
Gene Name: | NAD(P)H dehydrogenase, quinone 2 |
Gene Aliases: |
NMO2, Nmor2, Ox2 |
Location: |
Chr.13:33964655-33988465 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nqo2 | NM_001163239.1 | NP_001156711.1 | ||
NM_001163241.1 | NP_001156713.1 | |||
NM_020282.3 | NP_064678.1 | |||
AK008138.1 | BAB25486.1 | |||
AK008465.1 | BAB25683.1 | |||
AK010842.1 | BAB27217.1 | |||
AK049781.1 |
Target Gene Details
Entrez Gene ID: | 20719 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 6a |
Gene Aliases: |
4930482L21Rik, D330015H01Rik, PI-6, Serpinb6, Spi3, ovalbumin |
Location: |
Chr.13:33917918-34002794 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Serpinb6a | NM_001243192.1 | NP_001230121.1 | ||
CF732441.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |