Assay Details
Target Gene Details
Entrez Gene ID: | 69635 |
Gene Name: | death associated protein kinase 1 |
Gene Aliases: |
D13Ucla1, DAP-Kinase |
Location: |
Chr.13:60601947-60763191 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dapk1 | NM_001285917.1 | 21 | 2548 | NP_001272846.1 |
NM_029653.3 | 21 | 2548 | NP_083929.2 | |
NM_134062.2 | 21 | 2433 | NP_598823.1 | |
AK145922.1 | 21 | 2434 | BAE26753.1 | |
AY245540.2 | 21 | 2541 | AAO91934.2 | |
AY245541.1 | 21 | 2534 | AAO91935.1 | |
BC021490.1 | 19 | 2032 | AAH21490.1 | |
BQ745486.1 | 5 | 485 | ||
X97048.1 | 20 | 2383 | CAA65762.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)