Assay Details
Target Gene Details
Entrez Gene ID: | 80898 |
Gene Name: | endoplasmic reticulum aminopeptidase 1 |
Gene Aliases: |
Arts1, ERAAP, PILSA, PILSAP |
Location: |
Chr.13:74639624-74691931 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erap1 | NM_030711.4 | 7 | 1170 | NP_109636.1 |
AB047552.1 | 6 | 1158 | BAB11982.1 | |
AF227511.1 | 6 | 1146 | AAG44260.1 | |
AK029841.1 | 7 | 1169 | ||
AK030329.1 | 7 | 1171 | BAC26904.1 | |
AK040350.1 | 4 | 388 | BAC30569.1 | |
AK081181.1 | 7 | 1197 | BAC38159.1 | |
AK159406.1 | 7 | 1171 | ||
AK163954.1 | 7 | 1190 | ||
BC046610.1 | 7 | 1176 | AAH46610.1 |
More Information
Set Membership: |
Intragenic Exonic |