Assay Details
Target Gene Details
Entrez Gene ID: | 268663 |
Gene Name: | cadherin-related family member 2 |
Gene Aliases: |
Gm624, PC-LKC, Pcdh24 |
Location: |
Chr.13:54701463-54736662 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cdhr2 | NM_001033364.3 | 4 | 312 | NP_001028536.2 |
AK144400.1 | 4 | 313 | BAE25870.1 | |
BC141301.1 | 4 | 241 | ||
BC145624.1 | 4 | 241 |
More Information
Set Membership: |
Intragenic Exonic |