Assay Details
Target Gene Details
Entrez Gene ID: | 12380 |
Gene Name: | calpastatin |
Gene Aliases: |
- |
Location: |
Chr.13:74692366-74808874 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cast | NM_001301153.1 | NP_001288082.1 | ||
NM_001301155.1 | NP_001288084.1 | |||
NM_001301156.1 | NP_001288085.1 | |||
NM_001301157.1 | NP_001288086.1 | |||
NM_009817.4 | NP_033947.1 | |||
AB026997.1 | BAA84768.1 | |||
AK028841.1 | ||||
AK028843.1 | BAC26149.1 | |||
AK029293.1 | BAC26377.1 | |||
AK036576.1 | BAC29486.1 | |||
AK083310.1 | ||||
AK083395.1 | ||||
AK089108.1 | BAC40755.1 | |||
BC010603.1 | AAH10603.1 | |||
CN529927.1 | ||||
X62519.1 | CAA44385.1 |
More Information
Set Membership: |
Intragenic Non-exonic |