Assay Details
Target Gene Details
Entrez Gene ID: | 218268 |
Gene Name: | eukaryotic translation initiation factor 4E family member 1B |
Gene Aliases: |
Eif4eloo, Gm273 |
Location: |
Chr.13:54783927-54788459 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif4e1b | NM_001033269.3 | NP_001028441.1 | ||
NM_001039683.2 | NP_001034772.1 | |||
NM_001286178.1 | NP_001273107.1 | |||
NM_001286179.1 | NP_001273108.1 | |||
NM_001286180.1 | NP_001273109.1 | |||
NR_104406.1 | ||||
NR_104407.1 | ||||
AK139577.1 | BAE24071.1 | |||
BC139143.1 | ||||
BC139144.1 | ||||
DQ279473.1 | ABB88894.1 | |||
DQ279474.1 | ABB88895.1 | |||
DQ279478.1 | ABB88899.1 | |||
DQ279479.1 | ABB88900.1 | |||
DV653822.1 | ||||
DV659735.1 | ||||
DV662251.1 | ||||
DV662389.1 |
More Information
Set Membership: |
Intragenic Non-exonic |