Assay Details
Target Gene Details
Entrez Gene ID: | 74156 |
Gene Name: | acyl-CoA thioesterase 12 |
Gene Aliases: |
1300004O04Rik, 4930449F15Rik, AV027244, Cach, mCACH-1 |
Location: |
Chr.13:91741511-91786148 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acot12 | NM_028790.3 | NP_083066.1 | ||
AB078618.1 | BAB84021.1 | |||
AK004905.1 | BAB23658.1 | |||
AK033663.1 | ||||
AK034622.1 | BAC28775.1 | |||
BC025852.1 | AAH25852.1 | |||
BC132050.1 | ||||
BC137996.1 |
More Information
Set Membership: |
Intragenic Non-exonic |