Assay Details
Target Gene Details
Entrez Gene ID: | 373852 |
Gene Name: | RIKEN cDNA 4833422C13 gene |
Gene Aliases: |
- |
Location: |
Chr.13:91701665-91741872 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4833422C13Rik | NR_015501.1 | 1 | 243 | |
NR_028313.1 | 1 | 243 | ||
NR_028314.1 | 1 | 243 | ||
AK048302.1 | 1 | 244 | ||
AK135005.1 | 1 | 50 | ||
AK161222.1 | 1 | 185 | ||
AK162063.1 | 1 | 200 | BAE36704.1 | |
AK162854.1 | 1 | 156 |
Target Gene Details
Entrez Gene ID: | 74156 |
Gene Name: | acyl-CoA thioesterase 12 |
Gene Aliases: |
1300004O04Rik, 4930449F15Rik, AV027244, Cach, mCACH-1 |
Location: |
Chr.13:91741511-91786148 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Acot12 | NM_028790.3 | NP_083066.1 | ||
AB078618.1 | BAB84021.1 | |||
AK004905.1 | BAB23658.1 | |||
AK033663.1 | ||||
AK034622.1 | BAC28775.1 | |||
BC025852.1 | AAH25852.1 | |||
BC132050.1 | ||||
BC137996.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |