Assay Details
Target Gene Details
Entrez Gene ID: | 22632 |
Gene Name: | YY1 transcription factor |
Gene Aliases: |
AW488674, NF-E1 |
Location: |
Chr.12:108793269-108816632 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Yy1 | NM_009537.3 | NP_033563.2 | ||
AK076149.1 | BAC36219.1 | |||
AK166542.1 | BAE38842.1 | |||
BC055899.1 | AAH55899.1 | |||
M73963.1 | AAA40522.1 | |||
M74590.1 | AAA37521.1 |
More Information
Set Membership: |
Intragenic Non-exonic |