Assay Details
Target Gene Details
Entrez Gene ID: | 18755 |
Gene Name: | protein kinase C, eta |
Gene Aliases: |
Pkch |
Location: |
Chr.12:73585041-73778185 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prkch | NM_008856.3 | NP_032882.2 | ||
AK076352.1 | ||||
AK145817.1 | BAE26670.1 | |||
AK155951.1 | BAE33517.1 | |||
AK164044.1 | BAE37603.1 | |||
BC031121.1 | AAH31121.1 | |||
D90242.1 | BAA14288.1 |
More Information
Set Membership: |
Intragenic Non-exonic |