Assay Details
Target Gene Details
Entrez Gene ID: | 27260 |
Gene Name: | pleckstrin 2 |
Gene Aliases: |
- |
Location: |
Chr.12:78888691-78907182 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Plek2 | NM_013738.3 | NP_038766.1 | ||
AF157600.1 | AAD42973.1 | |||
AF170564.1 | AAD46924.1 | |||
BC028902.1 | AAH28902.1 |
More Information
Set Membership: |
Intragenic Non-exonic |