Assay Details
Target Gene Details
Entrez Gene ID: | 321017 |
Gene Name: | RIKEN cDNA 9230116L04 gene |
Gene Aliases: |
- |
Location: |
Chr.12:79088705-79103939 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
9230116L04Rik | NR_110486.1 | 3 | 750 | |
AK033827.1 | 3 | 751 |
Target Gene Details
Entrez Gene ID: | 110417 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class H |
Gene Aliases: |
2210416H01Rik, A930028P05Rik |
Location: |
Chr.12:79080664-79089754 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pigh | NM_029988.2 | 1 | 117 | NP_084264.1 |
AK020902.1 | 1 | 93 | ||
AK030744.1 | 1 | 81 | BAC27112.1 | |
AK167892.1 | 1 | 99 | BAE39904.1 | |
BC086806.1 | 1 | 68 | AAH86806.1 | |
BY737882.1 | 1 | 99 | ||
BY779412.1 | 1 | 117 |
More Information
Set Membership: |
Intragenic Exonic |