Assay Details
Target Gene Details
Entrez Gene ID: | 17252 |
Gene Name: | retinol dehydrogenase 11 |
Gene Aliases: |
2610319N22Rik, AI428145, AU045252, Arsdr1, C85936, CGI-82, HCBP12, M42C60, Mdt1, Psdr1, SCALD, UBE-1c1, Ube-1c |
Location: |
Chr.12:79174335-79191819 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rdh11 | NM_021557.5 | NP_067532.2 | ||
AB030503.1 | 1 | 343 | BAA82656.1 | |
AB035959.1 | BAA88521.1 | |||
AF474027.1 | 1 | 58 | AAL79910.1 | |
AK004413.1 | BAB23296.1 | |||
AK012056.1 | 1 | 40 | ||
AK135443.1 | 1 | 42 | BAE22534.1 | |
AY039032.1 | 1 | 486 | AAK91516.1 | |
BC018261.1 | 1 | 37 | AAH18261.1 |
More Information
Set Membership: |
Intragenic Non-exonic |