Assay Details
Target Gene Details
Entrez Gene ID: | 217944 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 5 |
Gene Aliases: |
4932413M22, C86120, D030051B22Rik, GFR, Mrgef, mKIAA0277, mmr-gef |
Location: |
Chr.12:117516479-117759737 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rapgef5 | NM_175930.5 | 7 | 1035 | NP_787126.3 |
AK029995.1 | 7 | 1036 | BAC26723.1 | |
AK030016.1 | 7 | 989 | BAC26736.1 | |
AK162220.1 | 5 | 668 | BAE36798.1 |
More Information
Set Membership: |
Intragenic Exonic |