Assay Details
Target Gene Details
Entrez Gene ID: | 217944 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 5 |
Gene Aliases: |
4932413M22, C86120, D030051B22Rik, GFR, Mrgef, mKIAA0277, mmr-gef |
Location: |
Chr.12:117516479-117759737 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rapgef5 | NM_175930.5 | 20 | 2270 | NP_787126.3 |
AK029995.1 | 20 | 2271 | BAC26723.1 | |
AK030016.1 | 20 | 2224 | BAC26736.1 | |
AK083591.1 | 10 | 1210 | BAC38963.1 | |
AK122234.1 | 8 | 1032 | BAC65516.1 | |
BC046627.1 | 8 | 991 | AAH46627.1 |
More Information
Set Membership: |
Intragenic Exonic |