Assay Details
Target Gene Details
Entrez Gene ID: | 17933 |
Gene Name: | myelin transcription factor 1-like |
Gene Aliases: |
2900046C06Rik, 2900093J19Rik, C630034G21Rik, Nztf1, Pmng1, Png-1, mKIAA1106 |
Location: |
Chr.12:29528311-29923216 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myt1l | NM_001093775.1 | NP_001087244.1 | ||
NM_001093776.1 | NP_001087245.1 | |||
NM_001093778.1 | NP_001087247.1 | |||
NM_008666.3 | NP_032692.2 | |||
AB093283.1 | BAC41467.1 | |||
AB212898.1 | BAE48255.1 | |||
AF004295.1 | AAC53457.1 | |||
AK012660.1 | ||||
AK036205.1 | ||||
AK049624.1 | ||||
BC094438.1 | ||||
BC131677.1 | ||||
BE653129.1 | ||||
U86338.1 | AAC53157.1 |
More Information
Set Membership: |
Intragenic Non-exonic |