Assay Details
Target Gene Details
Entrez Gene ID: | 56784 |
Gene Name: | Ral GTPase activating protein, alpha subunit 1 |
Gene Aliases: |
2310003F20Rik, 4930400K19Rik, AI563624, GRIPE, Garnl1, Tulip1, mKIAA0884 |
Location: |
Chr.12:55602890-55821575 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ralgapa1 | NM_001003719.2 | NP_001003719.1 | ||
NM_001112714.2 | NP_001106185.1 | |||
NM_001286263.1 | NP_001273192.1 | |||
NM_019994.5 | NP_064378.4 | |||
AY066011.1 | AAL47577.1 | |||
AY596972.1 | AAT49273.1 | |||
AY596973.1 | AAT49274.1 | |||
AY596974.1 | AAT49275.1 | |||
BC145119.1 | ||||
BC150707.1 | ||||
CN459409.1 |
More Information
Set Membership: |
Intragenic Non-exonic |