Assay Details
Target Gene Details
Entrez Gene ID: | 108156 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent), methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthase |
Gene Aliases: |
Dcs, E430024A07Rik, Mthfd |
Location: |
Chr.12:76255232-76319820 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mthfd1 | NM_138745.2 | NP_620084.2 | ||
AF364579.1 | AAL99692.1 | |||
AK042019.1 | BAC31133.2 | |||
AK042958.1 | BAC31419.1 | |||
AK088700.1 | BAC40513.1 | |||
AK129012.1 | ||||
AK133848.1 | BAE21884.1 | |||
AK159812.1 | BAE35392.1 | |||
AK165951.1 | ||||
BC008523.1 | AAH08523.1 |