Assay Details
Target Gene Details
Entrez Gene ID: | 320910 |
Gene Name: | integrin beta 8 |
Gene Aliases: |
4832412O06Rik |
Location: |
Chr.12:119158186-119238276 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb8 | NM_177290.3 | 3 | 784 | NP_796264.2 |
AK029281.1 | 3 | 1190 | ||
AK034644.1 | 3 | 1279 | ||
BC120691.1 | 3 | 784 | ||
BC125343.1 | 3 | 784 |
More Information
Set Membership: |
Intragenic Exonic |