Assay Details
Target Gene Details
Entrez Gene ID: | 192119 |
Gene Name: | dicer 1, ribonuclease type III |
Gene Aliases: |
1110006F08Rik, D12Ertd7e, mKIAA0928 |
Location: |
Chr.12:104687740-104751952 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dicer1 | NM_148948.2 | NP_683750.2 | ||
AB081470.1 | BAC15765.1 | |||
AF430845.1 | AAM21495.1 | |||
AK033198.1 | 1 | 1241 | ||
BC019805.1 | ||||
BC061198.1 | AAH61198.1 |