Assay Details
Target Gene Details
Entrez Gene ID: | 16997 |
Gene Name: | latent transforming growth factor beta binding protein 2 |
Gene Aliases: |
AW208642 |
Location: |
Chr.12:84783212-84877266 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ltbp2 | NM_013589.3 | 27 | 4208 | NP_038617.3 |
AF004874.1 | 28 | 4387 | AAB61611.1 | |
BC119785.1 | 27 | 4192 |
More Information
Set Membership: |
Intragenic Exonic |