Assay Details
Target Gene Details
Entrez Gene ID: | 70127 |
Gene Name: | D4, zinc and double PHD fingers, family 3 |
Gene Aliases: |
2810403B03Rik, 6530402L11Rik, BAF45C, C78788, CERD4, cer-d4 |
Location: |
Chr.12:83213751-83487736 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dpf3 | NM_001267625.1 | 3 | 281 | NP_001254554.1 |
NM_001267626.1 | 3 | 281 | NP_001254555.1 | |
NM_058212.2 | 3 | 281 | NP_478119.1 | |
AF362750.1 | 3 | 253 | AAK54538.1 | |
AK012970.1 | 3 | 286 | ||
AK039011.1 | 3 | 262 | BAC30204.1 | |
BC048572.1 | 3 | 161 | AAH48572.1 | |
FJ151218.1 | 3 | 253 |
More Information
Set Membership: |
Intragenic Exonic |