Assay Details
Target Gene Details
Entrez Gene ID: | 108156 |
Gene Name: | methylenetetrahydrofolate dehydrogenase (NADP+ dependent), methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthase |
Gene Aliases: |
Dcs, E430024A07Rik, Mthfd |
Location: |
Chr.12:76255232-76319820 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mthfd1 | NM_138745.2 | 11 | 1311 | NP_620084.2 |
AF364579.1 | 11 | 1092 | AAL99692.1 | |
AK042958.1 | 11 | 1147 | BAC31419.1 | |
AK088700.1 | 11 | 1108 | BAC40513.1 | |
AK129012.1 | 11 | 1245 | ||
AK133848.1 | 10 | 1046 | BAE21884.1 | |
AK159812.1 | 11 | 1123 | BAE35392.1 | |
AK165951.1 | 11 | 1143 | ||
AK213959.1 | 3 | 281 | ||
BC008523.1 | 11 | 1135 | AAH08523.1 |