Assay Details
Target Gene Details
Entrez Gene ID: | 217869 |
Gene Name: | eukaryotic translation initiation factor 5 |
Gene Aliases: |
2810011H21Rik, D12Ertd549e |
Location: |
Chr.12:111538101-111546753 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif5 | NM_173363.5 | NP_775539.1 | ||
NM_178041.2 | NP_829887.1 | |||
AK051980.1 | 2 | 730 | ||
AK076403.1 | BAC36325.1 | |||
AK159232.1 | BAE34918.1 | |||
AK160828.1 | ||||
AK160879.1 | 3 | 685 | ||
AK161554.1 | BAE36460.1 | |||
AK163368.1 | BAE37319.1 | |||
AK166775.1 | BAE39011.1 | |||
AK167331.1 | BAE39433.1 | |||
AK172048.1 | BAE42798.1 | |||
BC019769.1 | ||||
BC023074.1 | ||||
BC025523.1 | ||||
BC025557.1 | 2 | 345 | ||
BC029003.1 | ||||
BC039275.1 | AAH39275.1 | |||
BC056633.1 | AAH56633.1 |
More Information
Set Membership: |
Intragenic Non-exonic |