Assay Details
Target Gene Details
Entrez Gene ID: | 15519 |
Gene Name: | heat shock protein 90, alpha (cytosolic), class A member 1 |
Gene Aliases: |
86kDa, 89kDa, AL024080, AL024147, Hsp86-1, Hsp89, Hsp90, Hspca, hsp4 |
Location: |
Chr.12:110691036-110696395 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hsp90aa1 | NM_010480.5 | NP_034610.1 | ||
AK004658.1 | BAB23449.1 | |||
AK077095.1 | BAC36610.1 | |||
AK088975.1 | BAC40681.1 | |||
AK146944.1 | BAE27553.1 | |||
AK147641.1 | BAE28042.1 | |||
AK157849.1 | ||||
AK158753.1 | 1 | 97 | ||
AK161276.1 | BAE36287.1 | |||
AK166696.1 | BAE38952.1 | |||
AK166720.1 | BAE38969.1 | |||
AK166729.1 | BAE38976.1 | |||
AK166847.1 | BAE39066.1 | |||
AK166867.1 | BAE39080.1 | |||
AK166896.1 | ||||
AK167008.1 | BAE39185.1 | |||
AK167065.1 | BAE39226.1 | |||
AK167086.1 | BAE39243.1 | |||
AK167114.1 | BAE39262.1 | |||
AK167171.1 | BAE39308.1 | |||
AK167188.1 | BAE39319.1 | |||
AK168407.1 | ||||
AK168605.1 | BAE40472.1 | |||
AK168620.1 | BAE40484.1 | |||
AK183519.1 | ||||
BC018232.1 | ||||
BC046614.1 | AAH46614.1 | |||
BC049124.1 | AAH49124.2 | |||
BC094024.1 | AAH94024.1 | |||
M36830.1 | AAA37868.1 |
More Information
Set Membership: |
Intragenic Non-exonic |