Assay Details
Target Gene Details
Entrez Gene ID: | 18191 |
Gene Name: | neurexin III |
Gene Aliases: |
- |
Location: |
Chr.12:88722865-90334935 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrxn3 | NM_001198587.3 | 12 | 3591 | NP_001185516.2 |
NM_172544.3 | 11 | 2481 | NP_766132.2 | |
AK032126.1 | 11 | 2473 | BAC27716.1 | |
AK129205.1 | 10 | 2354 | BAC98015.2 | |
BC060719.1 | 13 | 3166 | AAH60719.1 |