Assay Details
Target Gene Details
Entrez Gene ID: | 319504 |
Gene Name: | neuronal cell adhesion molecule |
Gene Aliases: |
Bravo, C030017F07Rik, C130076O07Rik, mKIAA0343 |
Location: |
Chr.12:44328832-44605194 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrcam | NM_001146031.1 | 7 | 1001 | NP_001139503.1 |
NM_176930.4 | 7 | 1001 | NP_795904.3 | |
AJ543321.1 | 7 | 903 | CAD65848.1 | |
AK045259.1 | 6 | 702 | BAC32284.1 | |
AK048567.1 | 8 | 991 | BAC33377.1 | |
AK087693.1 | 7 | 953 | ||
AK122252.1 | 7 | 931 | BAC65534.1 | |
AK218057.1 | 2 | 80 |
More Information
Set Membership: |
Intragenic Exonic |