Assay Details
Target Gene Details
Entrez Gene ID: | 319504 |
Gene Name: | neuronal cell adhesion molecule |
Gene Aliases: |
Bravo, C030017F07Rik, C130076O07Rik, mKIAA0343 |
Location: |
Chr.12:44328832-44605194 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nrcam | NM_001146031.1 | NP_001139503.1 | ||
NM_176930.4 | NP_795904.3 | |||
AJ543321.1 | CAD65848.1 | |||
AK087693.1 | ||||
AK122252.1 | BAC65534.1 | |||
AK201445.1 |
More Information
Set Membership: |
Intragenic Non-exonic |