Assay Details
Target Gene Details
Entrez Gene ID: | 67216 |
Gene Name: | membrane bound O-acyltransferase domain containing 2 |
Gene Aliases: |
2810049G06Rik, AU022889, AW547221, LPCAT4, Moact2, Oact2 |
Location: |
Chr.12:24831487-24964400 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mboat2 | NM_001083341.1 | NP_001076810.1 | ||
NM_026037.3 | NP_080313.2 | |||
AB297383.1 | ||||
AB305046.1 | ||||
AK012931.1 | BAB28556.1 | |||
AK031682.1 | ||||
AK031824.1 | BAC27567.1 | |||
AK033106.1 | BAC28154.1 | |||
AK034873.1 | BAC28863.1 | |||
AK076045.1 | BAC36144.1 | |||
AK215458.1 | ||||
AU067514.1 | ||||
BC025020.1 | AAH25020.1 |
More Information
Set Membership: |
Intragenic Non-exonic |