Assay Details
Target Gene Details
Entrez Gene ID: | 70934 |
Gene Name: | RIKEN cDNA 4931403G20 gene |
Gene Aliases: |
- |
Location: |
Chr.12:69843686-69853671 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4931403G20Rik | NR_038172.1 | |||
NR_038173.1 | ||||
AK016427.1 |
Target Gene Details
Entrez Gene ID: | 399510 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 5 |
Gene Aliases: |
4432415E19Rik, GCKR, KHS, MAPKKKK5 |
Location: |
Chr.12:69803757-69893163 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Map4k5 | NM_201519.2 | 9 | 1011 | NP_958927.2 |
AK019468.1 | 9 | 858 | BAB31739.1 | |
AK032123.1 | 9 | 897 | ||
AK053775.1 | 9 | 1012 | BAC35517.1 | |
AK084891.1 | 9 | 719 | BAC39305.1 | |
AK195763.1 | 4 | 252 | ||
BC040381.1 | 9 | 860 | AAH40381.2 | |
BC048173.1 | 8 | 893 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |