Assay Details
Target Gene Details
Entrez Gene ID: | 399510 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 5 |
Gene Aliases: |
4432415E19Rik, GCKR, KHS, MAPKKKK5 |
Location: |
Chr.12:69803757-69893163 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Map4k5 | NM_201519.2 | NP_958927.2 | ||
AK019468.1 | BAB31739.1 | |||
AK032123.1 | ||||
AK045031.1 | BAC32190.1 | |||
AK053775.1 | BAC35517.1 | |||
AK084891.1 | BAC39305.1 | |||
BC040381.1 | AAH40381.2 | |||
BC048173.1 |