Assay Details
Target Gene Details
Entrez Gene ID: | 27274 |
Gene Name: | zinc finger protein 354B |
Gene Aliases: |
Kid2, Tcf17l1, Znf354b |
Location: |
Chr.11:50921786-50931635 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp354b | NM_013744.3 | NP_038772.1 | ||
AF184112.1 | AAF01034.1 | |||
AK148756.1 | ||||
BC107400.1 | AAI07401.1 | |||
BC107401.1 | AAI07402.1 |