Assay Details
Target Gene Details
Entrez Gene ID: | 27274 |
Gene Name: | zinc finger protein 354B |
Gene Aliases: |
Kid2, Tcf17l1, Znf354b |
Location: |
Chr.11:50921786-50931635 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp354b | NM_013744.3 | NP_038772.1 | ||
AK148756.1 |