Assay Details
Target Gene Details
Entrez Gene ID: | 207921 |
Gene Name: | family with sequence similarity 228, member B |
Gene Aliases: |
A830093I24Rik |
Location: |
Chr.12:4738397-4769267 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam228b | NM_175431.4 | NP_780640.2 | ||
AK044133.1 | BAC31793.1 | |||
AK050447.1 | BAC34259.1 | |||
BC100429.1 | AAI00430.1 | |||
BC145648.1 |