Assay Details
Target Gene Details
Entrez Gene ID: | 278507 |
Gene Name: | WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2 |
Gene Aliases: |
2610304F08Rik, AY100450, Gasp1, WFIKKNRP |
Location: |
Chr.11:94235951-94247793 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wfikkn2 | NM_181819.2 | 1 | 83 | NP_861540.2 |
AY100450.1 | 1 | 225 | AAM47508.1 | |
AY308804.1 | 1 | 53 | AAP72503.1 | |
BC113144.1 | 1 | 53 | AAI13145.1 | |
JQ080910.1 | 1 | 53 |