Assay Details
Target Gene Details
Entrez Gene ID: | 20926 |
Gene Name: | suppressor of Ty 6 |
Gene Aliases: |
5131400N11Rik, AI132449, SPT6, Supt6h, mKIAA0162 |
Location: |
Chr.11:78206749-78245703 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Supt6 | NM_009297.2 | NP_033323.2 | ||
AK129069.1 | BAC97879.1 | |||
AK170533.1 | ||||
BC072657.1 | AAH72657.1 | |||
U40375.1 | AAB18950.1 |
More Information
Set Membership: |
Intragenic Non-exonic |