Assay Details
Target Gene Details
Entrez Gene ID: | 104776 |
Gene Name: | aldehyde dehydrogenase family 6, subfamily A1 |
Gene Aliases: |
1110038I05Rik, AI314632, Mmsdh |
Location: |
Chr.12:84430717-84450950 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aldh6a1 | NM_134042.2 | NP_598803.1 | ||
AF297860.1 | AAG44988.1 | |||
AK033587.1 | BAC28375.1 | |||
AK082790.1 | ||||
AK128924.1 | ||||
AK147146.1 | BAE27715.1 | |||
AK155814.1 | BAE33445.1 | |||
AK164366.1 | ||||
AK169915.1 | BAE41455.1 | |||
AK170305.1 | BAE41702.1 | |||
AK171581.1 | BAE42539.1 | |||
AK171896.1 | BAE42726.1 | |||
AK183301.1 | ||||
AK197238.1 | ||||
AK212397.1 | ||||
AK217579.1 | ||||
AK219365.1 | ||||
BC031148.1 | AAH31148.1 | |||
BC033440.1 | AAH33440.1 | |||
BC115855.1 |
Target Gene Details
Entrez Gene ID: | 72873 |
Gene Name: | coiled-coil domain containing 176 |
Gene Aliases: |
2900006K08Rik, AI427784 |
Location: |
Chr.12:84409068-84441763 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccdc176 |
More Information
Set Membership: |
Intragenic Non-exonic |