Assay Details
Target Gene Details
Entrez Gene ID: | 11491 |
Gene Name: | a disintegrin and metallopeptidase domain 17 |
Gene Aliases: |
CD156b, Tace |
Location: |
Chr.12:21323509-21373632 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Adam17 | NM_001277266.1 | NP_001264195.1 | ||
NM_001291871.1 | NP_001278800.1 | |||
NM_009615.6 | NP_033745.4 | |||
NR_102380.1 | ||||
NR_120376.1 | ||||
AB021709.1 | BAA78578.1 | |||
AJ007365.1 | CAA07480.1 | |||
AK137693.1 | BAE23464.1 | |||
AK139471.1 | BAE24023.1 | |||
AK142441.1 | ||||
AK144161.1 | BAE25740.1 | |||
AK149288.1 | BAE28792.1 | |||
AK149614.1 | BAE28991.1 | |||
AK165739.1 | BAE38362.1 | |||
BC094655.1 | AAH94655.1 | |||
BC114999.1 | AAI15000.1 | |||
BC138416.1 | ||||
BC138420.1 | ||||
BC145270.1 | ||||
U69613.1 | AAD09627.1 | |||
U69614.1 | AAD09628.1 |
More Information
Set Membership: |
Intragenic Non-exonic |