Assay Details
Target Gene Details
Entrez Gene ID: | 70834 |
Gene Name: | sperm associated antigen 9 |
Gene Aliases: |
3110018C07Rik, 4733401I23Rik, 4831406C20Rik, AW552012, JLP, JSAP2, JSAP2a, Jip4, Mapk8ip4, syd1 |
Location: |
Chr.11:93996058-94126087 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 23 - Intron 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Spag9 | NM_001025428.1 | NP_001020599.1 | ||
NM_001025429.1 | NP_001020600.1 | |||
NM_001025430.1 | NP_001020601.1 | |||
NM_001199203.1 | NP_001186132.1 | |||
NM_001199204.1 | NP_001186133.1 | |||
NM_001199205.1 | NP_001186134.1 | |||
NM_027569.2 | NP_081845.2 | |||
AB047782.1 | BAD93176.1 | |||
AB367798.1 | ||||
AF327451.1 | AAN61564.1 | |||
AK049161.1 | BAC33577.1 | |||
AK147431.1 | BAE27907.1 | |||
AK147537.1 | BAE27980.1 | |||
AK172961.1 | BAD32239.1 | |||
AY823270.1 | AAX19462.1 | |||
BC060100.1 | AAH60100.1 | |||
BC060506.1 | AAH60506.1 | |||
BC094670.1 | AAH94670.1 | |||
FJ042496.1 |
More Information
Set Membership: |
Intragenic Non-exonic |