Assay Details
Target Gene Details
Entrez Gene ID: | 104771 |
Gene Name: | JNK1/MAPK8-associated membrane protein |
Gene Aliases: |
1200003C05Rik, 2310047L11Rik, Jamp |
Location: |
Chr.12:72085839-72101527 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Jkamp | NM_001205067.1 | NP_001191996.1 | ||
NM_024205.2 | NP_077167.1 | |||
AK004555.1 | BAC25088.1 | |||
BC002238.1 | AAH02238.1 |
More Information
Set Membership: |
Intragenic Non-exonic |